Product Details

SNP ID
rs61732847
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:170866492 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCCAATCACGAAGATTGTTCTTC[C/T]TAAGGTATTTCTTGGTAAGGTATTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RPL22L1 PubMed Links

Gene Details

Gene
RPL22L1
Gene Name
ribosomal protein L22 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099645.1 343 Missense Mutation AAG,AGG K86R NP_001093115.1
NM_001320451.1 343 Missense Mutation AAG,AGG K85R NP_001307380.1

View Full Product Details