Product Details

SNP ID
rs141199896
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:25201489 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTACAAGGTACTGGGCAGGGAGGG[A/G]GTCAGAGGGCCCAGGCTACTCCTGG
Phenotype
MIM: 123630
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRYBB3 PubMed Links

Gene Details

Gene
CRYBB3
Gene Name
crystallin beta B3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004076.4 Intron NP_004067.1
XM_011529902.2 Intron XP_011528204.1
XM_017028599.1 Intron XP_016884088.1
Gene
KIAA1671
Gene Name
KIAA1671
There are no transcripts associated with this gene.

View Full Product Details