Product Details

SNP ID
rs117025461
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51014792 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACATCTGGATCAGCTGGAGCGTAG[C/T]ATCTGGATCAGTTGGAGCGTATGAC
Phenotype
MIM: 602673 MIM: 604434 MIM: 605504
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLK10 PubMed Links

Gene Details

Gene
KLK10
Gene Name
kallikrein related peptidase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077500.1 944 UTR 3 NP_001070968.1
NM_002776.4 944 UTR 3 NP_002767.2
NM_145888.2 944 UTR 3 NP_665895.1
XM_005259061.3 944 UTR 3 XP_005259118.1
XM_005259062.3 944 UTR 3 XP_005259119.1
XM_006723287.3 944 UTR 3 XP_006723350.1
XM_006723289.3 944 UTR 3 XP_006723352.1
XM_017026993.1 944 UTR 3 XP_016882482.1
Gene
KLK11
Gene Name
kallikrein related peptidase 11
There are no transcripts associated with this gene.

Gene
KLK9
Gene Name
kallikrein related peptidase 9
There are no transcripts associated with this gene.

View Full Product Details