Product Details

SNP ID
rs35245338
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32129103 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCATAGCATCGGGCAGCTCCTTC[A/G]GGGTTCCCAGCTCGAAATAGTTCTG
Phenotype
MIM: 600984
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATF6B PubMed Links

Gene Details

Gene
ATF6B
Gene Name
activating transcription factor 6 beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136153.1 948 Intron NP_001129625.1
NM_004381.4 948 Intron NP_004372.3
Gene
FKBPL
Gene Name
FK506 binding protein like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022110.3 948 Silent Mutation CCC,CCT P226P NP_071393.2

View Full Product Details