Product Details

SNP ID
rs373645195
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:73213061 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAACACAAGGCTTCTTATACCTT[C/G]AGTAACTATATCTAGGGCAGTTAAT
Phenotype
MIM: 300026
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NAP1L2 PubMed Links

Gene Details

Gene
NAP1L2
Gene Name
nucleosome assembly protein 1 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021963.3 1814 UTR 3 NP_068798.1

View Full Product Details