Product Details

SNP ID
rs59733060
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5268440 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGCAGAAGGAGGGTGTCAGGGTC[A/G]CAGGAACACCTGCAAACTGGAAGAG
Phenotype
MIM: 142100
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HBE1 PubMed Links

Gene Details

Gene
HBE1
Gene Name
hemoglobin subunit epsilon 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005330.3 726 UTR 3 NP_005321.1

View Full Product Details