Product Details

SNP ID
rs61740746
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:70206462 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGAACTCAGACGCATCTACCTCC[A/G]AAGCGTCCTGATGGGCCGCTGCTTT
Phenotype
MIM: 602457
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FADD PubMed Links

Gene Details

Gene
FADD
Gene Name
Fas associated via death domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003824.3 913 Missense Mutation AAA,GAA K206E NP_003815.1

View Full Product Details