Product Details

SNP ID
rs36055795
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.6:28505744 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
ACCTTCTGTTCTTTTTCTCCATTCA[C/T]ATCCCCTTTCTCAAAGAGCTGGAAA
Phenotype
MIM: 607913
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPX6 PubMed Links

Gene Details

Gene
GPX6
Gene Name
glutathione peroxidase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182701.1 468 Missense Mutation ATG,GTG M140V NP_874360.1

View Full Product Details