Product Details

SNP ID
rs34427213
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:23137935 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAAGAGCTTTTCGGAGCCCACCA[A/G]TTGGTCCTGAATTGTTTCCTTTGCA
Phenotype
MIM: 603614
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TNFRSF10D PubMed Links

Gene Details

Gene
TNFRSF10D
Gene Name
TNF receptor superfamily member 10d
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003840.4 1191 Silent Mutation CTG,TTG L366L NP_003831.2

View Full Product Details