Product Details

SNP ID
rs34622674
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:23137998 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGTTGCCGAGGCATCCAGCAAGG[G/T]GCTGACTGAGAAGAGAGAAGAGATT
Phenotype
MIM: 603614
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TNFRSF10D PubMed Links

Gene Details

Gene
TNFRSF10D
Gene Name
TNF receptor superfamily member 10d
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003840.4 1128 Missense Mutation NP_003831.2

View Full Product Details