Product Details

SNP ID
rs35160621
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:1211585 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTGGCCAGGGTGGAGTGGGCGTC[A/G]GCCTGCTCCTCTTGGATGGGGGTCC
Phenotype
MIM: 614282 MIM: 603905 MIM: 600315
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SDF4 PubMed Links

Gene Details

Gene
SDF4
Gene Name
stromal cell derived factor 4
There are no transcripts associated with this gene.

Gene
TNFRSF18
Gene Name
TNF receptor superfamily member 18
There are no transcripts associated with this gene.

Gene
TNFRSF4
Gene Name
TNF receptor superfamily member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003327.3 1277 Silent Mutation GCC,GCT A268A NP_003318.1
XM_011542074.2 1277 Silent Mutation GCC,GCT A298A XP_011540376.1
XM_011542075.2 1277 Silent Mutation GCC,GCT A294A XP_011540377.1
XM_011542076.2 1277 Silent Mutation GCC,GCT A272A XP_011540378.1
XM_011542077.2 1277 Silent Mutation GCC,GCT A194A XP_011540379.1
XM_017002231.1 1277 Silent Mutation GCC,GCT A294A XP_016857720.1
XM_017002232.1 1277 Silent Mutation GCC,GCT A268A XP_016857721.1

View Full Product Details