Product Details
- SNP ID
-
rs116449103
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:179573166 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACATTTGGGACAGGTCTGGGGGCT[A/G]GAGGGACCAAAAAGAAGTTCTATAT
- Phenotype
-
MIM: 604958
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ACTL6A
PubMed Links
Gene Details
- Gene
- ACTL6A
- Gene Name
- actin like 6A
View Full Product Details