Product Details

SNP ID
hCV25642602
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:81116416 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTGATTAAATGGTCACAATTATT[C/T]TACCAGCTGTTGTAATAATTTATGC
Phenotype
MIM: 300385
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HMGN5 PubMed Links

Gene Details

Gene
HMGN5
Gene Name
high mobility group nucleosome binding domain 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030763.2 Intron NP_110390.1

View Full Product Details