Product Details

SNP ID
rs7674958
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:70389653 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGATTTGAAATTATTTACTTCTT[A/T]TTTCCACAGCCTGGTGAGAGTCAAA
Phenotype
MIM: 608936 MIM: 611593
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OPRPN PubMed Links

Gene Details

Gene
OPRPN
Gene Name
opiorphin prepropeptide
There are no transcripts associated with this gene.

Gene
SMR3B
Gene Name
submaxillary gland androgen regulated protein 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006685.3 Intron NP_006676.1

View Full Product Details