Product Details

SNP ID
rs6591699
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55838826 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGCTTCTCAGATTACCTGGAACT[G/T]CAAATTCCCCTCTTCTTTGTATTTC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR5D16 PubMed Links

Gene Details

Gene
OR5D16
Gene Name
olfactory receptor family 5 subfamily D member 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005496.1 75 Silent Mutation CTG,CTT L25L NP_001005496.1

View Full Product Details