Product Details

SNP ID
rs35572169
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:113878102 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCACACTATACTCTTCTGTTTGG[A/C]AAATCTGGCCTATGCATCTTTCTTT
Phenotype
MIM: 600917
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PPP1R3A PubMed Links

Gene Details

Gene
PPP1R3A
Gene Name
protein phosphatase 1 regulatory subunit 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002711.3 2567 Missense Mutation TGC,TTC C997F NP_002702.2
XM_005250473.3 2567 Missense Mutation TGC,TTC C796F XP_005250530.1

View Full Product Details