Product Details

SNP ID
rs35684337
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:225886736 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTACACTAGGCGCCTATGGCTG[C/G]AGCCTCCTTGGCACGAGCGCACCAT
Phenotype
MIM: 603037
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LEFTY1 PubMed Links

Gene Details

Gene
LEFTY1
Gene Name
left-right determination factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020997.3 1172 Silent Mutation CTC,CTG L364L NP_066277.1
Gene
TMEM63A
Gene Name
transmembrane protein 63A
There are no transcripts associated with this gene.

View Full Product Details