Product Details

SNP ID
rs77577869
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:97927260 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAATGTTATTTCTTTCAGATATG[A/G]TCTACAAATAGAAAAAATAATATTA
Phenotype
MIM: 610715
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HEMGN PubMed Links

Gene Details

Gene
HEMGN
Gene Name
hemogen
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018437.4 1744 UTR 3 NP_060907.2
NM_197978.2 1744 UTR 3 NP_932095.1
XM_005252086.1 1744 UTR 3 XP_005252143.1
XM_011518845.2 1744 UTR 3 XP_011517147.1
XM_011518846.2 1744 UTR 3 XP_011517148.1
Gene
TRMO
Gene Name
tRNA methyltransferase O
There are no transcripts associated with this gene.

View Full Product Details