Product Details

SNP ID
rs3136717
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:42338923 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGAGGAAACGGGTGGTCACTGTCT[C/T]TTAGAGGCTGCCATATCCCCTTCCA
Phenotype
MIM: 603258 MIM: 174760
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IKBKB PubMed Links

Gene Details

Gene
IKBKB
Gene Name
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta
There are no transcripts associated with this gene.

Gene
POLB
Gene Name
DNA polymerase beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002690.2 Intron NP_002681.1
XM_005273535.3 Intron XP_005273592.1
XM_005273536.3 Intron XP_005273593.1
XM_005273537.3 Intron XP_005273594.1
XM_005273538.2 Intron XP_005273595.1
XM_005273539.2 Intron XP_005273596.1
XM_005273540.4 Intron XP_005273597.1
XM_006716353.2 Intron XP_006716416.1
XM_017013583.1 Intron XP_016869072.1
XM_017013584.1 Intron XP_016869073.1

View Full Product Details