Product Details

SNP ID
rs3737277
Assay Type
Validated
NCBI dbSNP Submissions
78
Location
Chr.1:247860664 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
AACTTATGAGGAAAGAGTTGGAGGA[T/C]GCCTTGACTCAGGAGGCCAACGTGG
Phenotype
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
TRIM58 PubMed Links

Gene Details

Gene
TRIM58
Gene Name
tripartite motif containing 58
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015431.3 516 Silent Mutation GAC,GAT D156D NP_056246.3

View Full Product Details