Product Details

SNP ID
rs7890400
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.X:24060240 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATGTGGAACAAATCCAAGATGAAAT[A/G]TTTAAAGGGGAACTAAGGCCTTTAA
Phenotype
MIM: 300161
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EIF2S3 PubMed Links

Gene Details

Gene
EIF2S3
Gene Name
eukaryotic translation initiation factor 2 subunit gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001415.3 Intron NP_001406.1

View Full Product Details