Product Details

SNP ID
rs16982874
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:24060277 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTAAGGCCTTTAAGGATCATATCT[A/G]TTACCTAAGGGTGACATGAATGGGA
Phenotype
MIM: 300161
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EIF2S3 PubMed Links

Gene Details

Gene
EIF2S3
Gene Name
eukaryotic translation initiation factor 2 subunit gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001415.3 Intron NP_001406.1

View Full Product Details