Product Details

SNP ID
rs35300823
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50770705 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAAGATGAACTCTTCCGGATGCCT[A/G]TCTGAGGAGGTGGGGTCCCTCCGCC
Phenotype
MIM: 603195
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR32 PubMed Links

Gene Details

Gene
GPR32
Gene Name
G protein-coupled receptor 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001506.2 242 Silent Mutation CTA,CTG L35L NP_001497.1
Gene
LOC105372440
Gene Name
uncharacterized LOC105372440
There are no transcripts associated with this gene.

View Full Product Details