Product Details

SNP ID
rs72551392
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.4:68670110 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
GCCAACAGAGAATCGAAGACTGTAC[A/G]GAAAGGGTATGTTAAATAGTTCAGC
Phenotype
MIM: 600069
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UGT2B15 PubMed Links

Gene Details

Gene
UGT2B15
Gene Name
UDP glucuronosyltransferase family 2 member B15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001076.3 667 Missense Mutation CCG,CTG P170L NP_001067.2

View Full Product Details