Product Details

SNP ID
rs34878463
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81869746 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCGCAGCCCAGACTCACCTGCG[A/G]AAACGGCCACGCGGCCCAGCAGGGC
Phenotype
MIM: 601925 MIM: 176790
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGDIA PubMed Links

Gene Details

Gene
ARHGDIA
Gene Name
Rho GDP dissociation inhibitor alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185077.2 546 Missense Mutation TCC,TTC S62F NP_001172006.1
NM_001185078.2 546 Missense Mutation TCC,TTC S62F NP_001172007.1
NM_001301240.1 546 Missense Mutation TCC,TTC S62F NP_001288169.1
NM_001301241.1 546 Missense Mutation TCC,TTC S62F NP_001288170.1
NM_001301242.1 546 Missense Mutation TCC,TTC S62F NP_001288171.1
NM_001301243.1 546 Missense Mutation TCC,TTC S62F NP_001288172.1
NM_004309.5 546 Missense Mutation TCC,TTC S62F NP_004300.1
XM_011523574.1 546 Missense Mutation TCC,TTC S62F XP_011521876.1
Gene
P4HB
Gene Name
prolyl 4-hydroxylase subunit beta
There are no transcripts associated with this gene.

View Full Product Details