Product Details

SNP ID
hCV25650531
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:75404032 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCTTCTTTCTCTGTCTCCCACTC[C/T]GCACGGGTACCGAGAACTTTCCGGG
Phenotype
MIM: 613902
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101929234 PubMed Links

Gene Details

Gene
LOC101929234
Gene Name
uncharacterized LOC101929234
There are no transcripts associated with this gene.

Gene
ZNF503
Gene Name
zinc finger protein 503
There are no transcripts associated with this gene.

Gene
ZNF503-AS2
Gene Name
ZNF503 antisense RNA 2
There are no transcripts associated with this gene.

View Full Product Details