Product Details

SNP ID
rs7090298
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.10:75403012 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GGTACCCGAGACCAGCGGCGGCAAC[G/T]TCTAACACGGGAGATTTCCCGCCAC
Phenotype
MIM: 613902
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC101929234 PubMed Links

Gene Details

Gene
LOC101929234
Gene Name
uncharacterized LOC101929234
There are no transcripts associated with this gene.

Gene
ZNF503
Gene Name
zinc finger protein 503
There are no transcripts associated with this gene.

Gene
ZNF503-AS2
Gene Name
ZNF503 antisense RNA 2
There are no transcripts associated with this gene.

View Full Product Details