Product Details

SNP ID
rs377033484
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33251840 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTCCCTTACCACTGATCCCATCA[C/T]TACCATATTTTCCTCATACCTTCTT
Phenotype
MIM: 603443
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HCG25 PubMed Links

Gene Details

Gene
HCG25
Gene Name
HLA complex group 25 (non-protein coding)
There are no transcripts associated with this gene.

Gene
VPS52
Gene Name
VPS52, GARP complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289174.1 Intron NP_001276103.1
NM_001289175.1 Intron NP_001276104.1
NM_001289176.1 Intron NP_001276105.1
NM_022553.5 Intron NP_072047.4
XM_011514797.1 Intron XP_011513099.1
XM_011514798.2 Intron XP_011513100.1
XM_011514799.1 Intron XP_011513101.1
XM_017011177.1 Intron XP_016866666.1
XM_017011178.1 Intron XP_016866667.1
XM_017011179.1 Intron XP_016866668.1

View Full Product Details