Product Details

SNP ID
hCV25652910
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:150999906 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCACTGGAGTGTTTGCAGGTGATG[A/G]TCCTCTCCACCAGCTGGTTGTAACT
Phenotype
MIM: 603319
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANXA9 PubMed Links

Gene Details

Gene
ANXA9
Gene Name
annexin A9
There are no transcripts associated with this gene.

Gene
FAM63A
Gene Name
family with sequence similarity 63 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040217.2 2478 Missense Mutation ACC,ATC T123I NP_001035307.1
NM_001163258.1 2478 Missense Mutation ACC,ATC T313I NP_001156730.1
NM_001163259.1 2478 Missense Mutation ACC,ATC T170I NP_001156731.1
NM_001163260.1 2478 Missense Mutation ACC,ATC T123I NP_001156732.1
NM_001319998.1 2478 Missense Mutation ACC,ATC T265I NP_001306927.1
NM_018379.4 2478 Missense Mutation ACC,ATC T265I NP_060849.2
Gene
PRUNE1
Gene Name
prune exopolyphosphatase
There are no transcripts associated with this gene.

View Full Product Details