Product Details

SNP ID
rs61736363
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30185263 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTCCGCCACGTGTTTGGACAGCC[G/C]GCCAAGGCCGACCAGTGCTATGAAG
Phenotype
MIM: 605000 MIM: 615822
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
BOLA2B PubMed Links

Gene Details

Gene
BOLA2B
Gene Name
bolA family member 2B
There are no transcripts associated with this gene.

Gene
CORO1A
Gene Name
coronin 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193333.2 489 Silent Mutation CCC,CCG P18P NP_001180262.1
NM_007074.3 489 Silent Mutation CCC,CCG P18P NP_009005.1
XM_011545714.2 489 Silent Mutation CCC,CCG P18P XP_011544016.1
XM_017022885.1 489 Silent Mutation CCC,CCG P18P XP_016878374.1
XM_017022886.1 489 Silent Mutation CCC,CCG P18P XP_016878375.1
Gene
SLX1A
Gene Name
SLX1 homolog A, structure-specific endonuclease subunit
There are no transcripts associated with this gene.

Gene
SLX1A-SULT1A3
Gene Name
SLX1A-SULT1A3 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details