Product Details

SNP ID
rs55808472
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:24920370 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGAAGTTTCAGGGTCAGCCCAT[A/G]CACACAGGGTCCCTTGCGAGAGGCC
Phenotype
MIM: 608293 MIM: 610238
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP17 PubMed Links

Gene Details

Gene
ARHGAP17
Gene Name
Rho GTPase activating protein 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006634.2 Intron NP_001006635.1
NM_018054.5 Intron NP_060524.4
XM_011545873.2 Intron XP_011544175.1
XM_011545874.2 Intron XP_011544176.1
XM_011545875.2 Intron XP_011544177.1
XM_011545876.2 Intron XP_011544178.1
XM_011545877.2 Intron XP_011544179.1
XM_011545878.2 Intron XP_011544180.1
XM_017023390.1 Intron XP_016878879.1
Gene
SLC5A11
Gene Name
solute carrier family 5 member 11
There are no transcripts associated with this gene.

View Full Product Details