Product Details

SNP ID
rs74575755
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:31325077 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAGCACAAAAAGCAGTGCCCTGC[A/G]GAGTTCACTGACTCCCGCGGCGTAC
Phenotype
MIM: 615027
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM60A PubMed Links

Gene Details

Gene
FAM60A
Gene Name
family with sequence similarity 60 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135811.1 Intron NP_001129283.1
NM_001135812.1 Intron NP_001129284.1
NM_021238.2 Intron NP_067061.1
XM_011520803.2 Intron XP_011519105.1
XM_017019758.1 Intron XP_016875247.1
XM_017019759.1 Intron XP_016875248.1
XM_017019760.1 Intron XP_016875249.1
XM_017019761.1 Intron XP_016875250.1
XM_017019762.1 Intron XP_016875251.1
Gene
FLJ13224
Gene Name
uncharacterized LOC79857
There are no transcripts associated with this gene.

View Full Product Details