Product Details

SNP ID
rs7257699
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:843454 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTCCCTGACGCCTGGACTCCCCC[A/C]CTGCAGGTGCTGCCCGAGCTGCGGA
Phenotype
MIM: 130130 MIM: 177020
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ELANE PubMed Links

Gene Details

Gene
ELANE
Gene Name
elastase, neutrophil expressed
There are no transcripts associated with this gene.

Gene
PRTN3
Gene Name
proteinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002777.3 Intron NP_002768.3
XM_011528136.1 Intron XP_011526438.1

View Full Product Details