Product Details

SNP ID
rs7724858
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:1225588 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAGCTGCACTGGGTTTGCCGTGGG[A/G]CTGGGGAACATTTGGCGGTTCCCAT
Phenotype
MIM: 610300 MIM: 608893
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC6A18 PubMed Links

Gene Details

Gene
SLC6A18
Gene Name
solute carrier family 6 member 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182632.2 234 Silent Mutation GGA,GGG G37G NP_872438.2
Gene
SLC6A19
Gene Name
solute carrier family 6 member 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003841.2 234 Intron NP_001003841.1

View Full Product Details