Product Details

SNP ID
hCV25742946
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:73500614 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACCCTTGGAGGCCCCTGAAAAGG[G/T]ACCAAATTCTGGCATATAATATTAG
Phenotype
MIM: 114106
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PPP3CB PubMed Links

Gene Details

Gene
PPP3CB
Gene Name
protein phosphatase 3 catalytic subunit beta
There are no transcripts associated with this gene.

Gene
PPP3CB-AS1
Gene Name
PPP3CB antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
USP54
Gene Name
ubiquitin specific peptidase 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320437.1 Intron NP_001307366.1
NM_001320441.1 Intron NP_001307370.1
NM_152586.3 Intron NP_689799.3

View Full Product Details