Product Details

SNP ID
hCV25743407
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:162374736 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCGGTTACCTCCACCCCACCAC[A/G]ATTGCAGTTCTGAGAGCAGACGCAA
Phenotype
MIM: 605551
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf111 PubMed Links

Gene Details

Gene
C1orf111
Gene Name
chromosome 1 open reading frame 111
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182581.3 Intron NP_872387.2
Gene
C1orf226
Gene Name
chromosome 1 open reading frame 226
There are no transcripts associated with this gene.

Gene
NOS1AP
Gene Name
nitric oxide synthase 1 adaptor protein
There are no transcripts associated with this gene.

View Full Product Details