Product Details
- SNP ID
-
rs9379860
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:26370377 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGGGTTATGAGGATGGAGGGATCCA[C/T]CTGGAGTGCAGGTCCACCGGCTGGT
- Phenotype
-
MIM: 613594
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
BTN3A2
PubMed Links
Gene Details
- Gene
- BTN3A2
- Gene Name
- butyrophilin subfamily 3 member A2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001197246.2 |
737 |
Silent Mutation |
CAC,CAT |
H163H |
NP_001184175.1 |
NM_001197247.2 |
737 |
Silent Mutation |
CAC,CAT |
H163H |
NP_001184176.1 |
NM_001197248.2 |
737 |
Silent Mutation |
CAC,CAT |
H140H |
NP_001184177.1 |
NM_001197249.2 |
737 |
Silent Mutation |
CAC,CAT |
H121H |
NP_001184178.1 |
NM_007047.4 |
737 |
Silent Mutation |
CAC,CAT |
H163H |
NP_008978.2 |
XM_005248827.3 |
737 |
Silent Mutation |
CAC,CAT |
H163H |
XP_005248884.1 |
XM_005248831.3 |
737 |
Silent Mutation |
CAC,CAT |
H163H |
XP_005248888.1 |
XM_005248832.3 |
737 |
Silent Mutation |
CAC,CAT |
H163H |
XP_005248889.1 |
XM_006714979.3 |
737 |
Silent Mutation |
CAC,CAT |
H163H |
XP_006715042.1 |
XM_006714980.3 |
737 |
Silent Mutation |
CAC,CAT |
H163H |
XP_006715043.1 |
XM_006714981.3 |
737 |
Silent Mutation |
CAC,CAT |
H163H |
XP_006715044.1 |
XM_006714982.3 |
737 |
Silent Mutation |
CAC,CAT |
H121H |
XP_006715045.1 |
XM_011514267.2 |
737 |
Silent Mutation |
CAC,CAT |
H163H |
XP_011512569.1 |
XM_011514268.2 |
737 |
Silent Mutation |
CAC,CAT |
H163H |
XP_011512570.1 |
XM_011514269.2 |
737 |
Silent Mutation |
CAC,CAT |
H121H |
XP_011512571.1 |
XM_011514270.2 |
737 |
Intron |
|
|
XP_011512572.1 |
XM_011514271.2 |
737 |
Intron |
|
|
XP_011512573.1 |
XM_017010211.1 |
737 |
Silent Mutation |
CAC,CAT |
H163H |
XP_016865700.1 |
XM_017010212.1 |
737 |
Silent Mutation |
CAC,CAT |
H163H |
XP_016865701.1 |
XM_017010213.1 |
737 |
Silent Mutation |
CAC,CAT |
H121H |
XP_016865702.1 |
XM_017010214.1 |
737 |
Silent Mutation |
CAC,CAT |
H121H |
XP_016865703.1 |
XM_017010215.1 |
737 |
Intron |
|
|
XP_016865704.1 |
XM_017010216.1 |
737 |
Silent Mutation |
CAC,CAT |
H121H |
XP_016865705.1 |
XM_017010217.1 |
737 |
Intron |
|
|
XP_016865706.1 |
View Full Product Details