Product Details

SNP ID
rs117030628
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:16439029 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTTAAGGGCCTGGCGCACATAA[G/T]GTGTGACCTTTTCATTCCCGTTGTT
Phenotype
MIM: 606676
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LRRC75A PubMed Links

Gene Details

Gene
LRRC75A
Gene Name
leucine rich repeat containing 75A
There are no transcripts associated with this gene.

Gene
LRRC75A-AS1
Gene Name
LRRC75A antisense RNA 1
There are no transcripts associated with this gene.

Gene
SNORD49A
Gene Name
small nucleolar RNA, C/D box 49A
There are no transcripts associated with this gene.

Gene
SNORD49B
Gene Name
small nucleolar RNA, C/D box 49B
There are no transcripts associated with this gene.

Gene
SNORD65
Gene Name
small nucleolar RNA, C/D box 65
There are no transcripts associated with this gene.

Gene
TRPV2
Gene Name
transient receptor potential cation channel subfamily V member 2
There are no transcripts associated with this gene.

View Full Product Details