Product Details

SNP ID
rs542216976
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:37005690 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGAGCCGACCTTCTGAGCCCTCA[A/G]GAAAGATCAGAACAGATTCATGGGT
Phenotype
MIM: 605091
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FGD2 PubMed Links

Gene Details

Gene
FGD2
Gene Name
FYVE, RhoGEF and PH domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173558.3 36 UTR 5 NP_775829.2
XM_011514372.2 36 UTR 5 XP_011512674.1
XM_011514373.2 36 UTR 5 XP_011512675.1
XM_017010429.1 36 UTR 5 XP_016865918.1

View Full Product Details