Product Details

SNP ID
rs34729789
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:44148853 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGGTCCAGTAGTCAATGTATTTC[A/G]CAGGAATAGGTGATGGCATACCAAG
Phenotype
MIM: 300817
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EFHC2 PubMed Links

Gene Details

Gene
EFHC2
Gene Name
EF-hand domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025184.3 1995 Missense Mutation GCG,GTG A731V NP_079460.2
XM_006724562.2 1995 Missense Mutation GCG,GTG A535V XP_006724625.1

View Full Product Details