Product Details

SNP ID
rs112057932
Assay Type
Functionally tested
NCBI dbSNP Submissions
22
Location
Chr.1:89132450 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTGAAAATCCCCAATTTTTAAG[A/G]GACCGAGGTTTGTATTTAACAATTT
Phenotype
MIM: 600412 MIM: 612468
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GBP2 PubMed Links

Gene Details

Gene
GBP2
Gene Name
guanylate binding protein 2
There are no transcripts associated with this gene.

Gene
GBP7
Gene Name
guanylate binding protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207398.2 Intron NP_997281.2

View Full Product Details