Product Details

SNP ID
rs62620661
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57222989 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGTTCCTGTCCCCAGTGACACTG[C/T]GTGTGTACACATGGGTGTGGGCAAA
Phenotype
MIM: 107770 MIM: 604637 MIM: 138450
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRP1 PubMed Links

Gene Details

Gene
LRP1
Gene Name
LDL receptor related protein 1
There are no transcripts associated with this gene.

Gene
NXPH4
Gene Name
neurexophilin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007224.3 Intron NP_009155.1
XM_017018747.1 Intron XP_016874236.1
Gene
SHMT2
Gene Name
serine hydroxymethyltransferase 2
There are no transcripts associated with this gene.

View Full Product Details