Product Details

SNP ID
rs8106673
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51481471 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGGATTCTGTGGCTGCTGACTGCC[A/T]CACAAATGTCACCAACATCACGTGG
Phenotype
MIM: 606094
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
CEACAM18 PubMed Links
Additional Information
For this assay, SNP(s) [rs12610545] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CEACAM18
Gene Name
carcinoembryonic antigen related cell adhesion molecule 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278392.1 479 Missense Mutation CAC,CTC H160L NP_001265321.1
XM_011527258.2 479 Missense Mutation CAC,CTC H160L XP_011525560.1
Gene
SIGLEC12
Gene Name
sialic acid binding Ig like lectin 12 (gene/pseudogene)
There are no transcripts associated with this gene.

View Full Product Details