Product Details

SNP ID
rs41307894
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:37567616 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGACCATTTGTTTCAGTGGGTCTT[C/G]ACAACTGGAGTTCTAGTTACTCTAC
Phenotype
MIM: 602135 MIM: 609365
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DNALI1 PubMed Links

Gene Details

Gene
DNALI1
Gene Name
dynein axonemal light intermediate chain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003462.3 Intron NP_003453.2
XM_005271172.2 Intron XP_005271229.1
Gene
GNL2
Gene Name
G protein nucleolar 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323623.1 Intron NP_001310552.1
NM_001323624.1 Intron NP_001310553.1
NM_013285.2 Intron NP_037417.1

View Full Product Details