Product Details

SNP ID
rs76505373
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:86352143 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAAAATCATTTAACTCTTGAATC[C/T]TTTAGGTTTTCAACTTTTTAGACAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR7856 PubMed Links

Gene Details

Gene
MIR7856
Gene Name
microRNA 7856
There are no transcripts associated with this gene.

Gene
ODF2L
Gene Name
outer dense fiber of sperm tails 2 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007022.2 2280 UTR 3 NP_001007023.2
NM_001184765.1 2280 Intron NP_001171694.1
NM_001184766.1 2280 Intron NP_001171695.1
NM_020729.2 2280 UTR 3 NP_065780.2
XM_005271054.3 2280 Intron XP_005271111.1
XM_011541815.2 2280 Intron XP_011540117.1
XM_011541819.2 2280 UTR 3 XP_011540121.1
XM_011541821.2 2280 UTR 3 XP_011540123.1
XM_011541823.2 2280 Intron XP_011540125.1
XM_011541824.2 2280 Intron XP_011540126.1
XM_017001876.1 2280 Intron XP_016857365.1
XM_017001877.1 2280 Intron XP_016857366.1
XM_017001878.1 2280 Intron XP_016857367.1
XM_017001879.1 2280 UTR 3 XP_016857368.1
XM_017001880.1 2280 Intron XP_016857369.1
XM_017001881.1 2280 Intron XP_016857370.1
XM_017001882.1 2280 Intron XP_016857371.1

View Full Product Details