Product Details

SNP ID
rs116213822
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.13:27271579 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGCTCCCTGCTTTTATGCTTGCG[G/T]GTTTCTTTTTACGGATGGGAGTTTG
Phenotype
MIM: 612403
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
RASL11A PubMed Links

Gene Details

Gene
RASL11A
Gene Name
RAS like family 11 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_206827.1 Intron NP_996563.1

View Full Product Details