Product Details

SNP ID
rs17110961
Assay Type
Functionally tested
NCBI dbSNP Submissions
28
Location
Chr.1:54717428 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATAGTACATTATTACATGATTTAG[A/G]ATCTATTTTTAATGAGCCTTTTAAA
Phenotype
MIM: 606753
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MROH7 PubMed Links

Gene Details

Gene
MROH7
Gene Name
maestro heat like repeat family member 7
There are no transcripts associated with this gene.

Gene
MROH7-TTC4
Gene Name
MROH7-TTC4 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
TTC4
Gene Name
tetratricopeptide repeat domain 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291333.1 Intron NP_001278262.1
NM_004623.4 Intron NP_004614.3

View Full Product Details