Product Details
- SNP ID
-
hCV25753328
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:27271463 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGCTTGTTCTACTCCTTCGGTTG[A/G]TTTTCCTATTTCCTTTTCAGCAATG
- Phenotype
-
MIM: 612403
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RASL11A
PubMed Links
Gene Details
- Gene
- RASL11A
- Gene Name
- RAS like family 11 member A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_206827.1 |
|
Intron |
|
|
NP_996563.1 |
View Full Product Details