Product Details

SNP ID
rs564694891
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:47778997 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGAATGCAGACTTGAATGTTGTA[C/T]ACATCATTTACAGTTACAAAAATCG
Phenotype
MIM: 607614
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NUP160 PubMed Links

Gene Details

Gene
NUP160
Gene Name
nucleoporin 160
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318399.1 4543 Intron NP_001305328.1
NM_015231.2 4543 UTR 3 NP_056046.1

View Full Product Details