Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001005290.3 | 1074 | Silent Mutation | NP_001005290.1 | ||
NM_001032291.2 | 1074 | Missense Mutation | NP_001027462.1 | ||
NM_032636.7 | 1074 | Missense Mutation | NP_116025.1 | ||
XM_005271283.2 | 1074 | Missense Mutation | XP_005271340.1 | ||
XM_011542306.2 | 1074 | Missense Mutation | XP_011540608.1 | ||
XM_017002560.1 | 1074 | Missense Mutation | XP_016858049.1 | ||
XM_017002561.1 | 1074 | Missense Mutation | XP_016858050.1 | ||
XM_017002562.1 | 1074 | Missense Mutation | XP_016858051.1 | ||
XM_017002563.1 | 1074 | Missense Mutation | XP_016858052.1 | ||
XM_017002564.1 | 1074 | Missense Mutation | XP_016858053.1 | ||
XM_017002565.1 | 1074 | Missense Mutation | XP_016858054.1 | ||
XM_017002566.1 | 1074 | Missense Mutation | XP_016858055.1 | ||
XM_017002567.1 | 1074 | Missense Mutation | XP_016858056.1 | ||
XM_017002568.1 | 1074 | Missense Mutation | XP_016858057.1 | ||
XM_017002569.1 | 1074 | Missense Mutation | XP_016858058.1 | ||
XM_017002570.1 | 1074 | Missense Mutation | XP_016858059.1 | ||
XM_017002571.1 | 1074 | Missense Mutation | XP_016858060.1 | ||
XM_017002572.1 | 1074 | Missense Mutation | XP_016858061.1 | ||
XM_017002573.1 | 1074 | Missense Mutation | XP_016858062.1 | ||
XM_017002574.1 | 1074 | Missense Mutation | XP_016858063.1 | ||
XM_017002575.1 | 1074 | Missense Mutation | XP_016858064.1 | ||
XM_017002576.1 | 1074 | Silent Mutation | XP_016858065.1 | ||
XM_017002577.1 | 1074 | Silent Mutation | XP_016858066.1 |